siRNA Details
Gene Symbol: P2ry6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057124.2 | NP_476465.1 | 3 | 276 | ||
XM_006229711.2 | XP_006229773.1 | 2 | 360 | |||
XM_006229712.2 | XP_006229774.1 | 3 | 1712 | |||
XM_006229713.2 | XP_006229775.1 | 3 | 429 | |||
XM_006229714.2 | XP_006229776.1 | 3 | 1376 | |||
XM_006229715.2 | XP_006229777.1 | 4 | 1859 | |||
XM_006229716.2 | XP_006229778.1 | 3 | 1097 | |||
XM_006229717.2 | XP_006229779.1 | 3 | 3667 | |||
GenBank mRNA | BC072520.1 | - | 3 | 276 | 7105972 | |
D63665.1 | - | 4 | 500 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Panther Classification:
