Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TCF7L2 SIRNA Assays ›
Gene Symbol:
TCF7L2
Gene Name:
transcription factor 7 like 2
Gene Aliases:
TCF-4, TCF4
Chromosome Location:
Chr.10: 112950219 - 113167678 on Build GRCh38
UniGene ID:
Hs.593995
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s13881
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TCF7L2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001146274.1
NP_001139746.1
1
662
NM_001146283.1
NP_001139755.1
1
662
NM_001146284.1
NP_001139756.1
1
662
NM_001146285.1
NP_001139757.1
1
662
NM_001146286.1
NP_001139758.1
1
662
NM_001198525.1
NP_001185454.1
1
662
NM_001198526.1
NP_001185455.1
1
662
NM_001198527.1
NP_001185456.1
1
662
NM_001198528.1
NP_001185457.1
1
662
NM_001198529.1
NP_001185458.1
1
662
NM_001198530.1
NP_001185459.1
1
662
NM_001198531.1
NP_001185460.1
1
662
NM_030756.4
NP_110383.2
1
662
XM_005270071.1
XP_005270128.1
1
693
XM_005270075.1
XP_005270132.1
1
693
XM_005270077.1
XP_005270134.1
1
693
XM_005270078.1
XP_005270135.1
1
693
XM_005270079.1
XP_005270136.1
1
693
XM_005270080.1
XP_005270137.1
1
693
XM_005270084.1
XP_005270141.1
1
693
XM_005270085.1
XP_005270142.1
1
693
XM_005270086.1
XP_005270143.1
1
693
XM_005270088.1
XP_005270145.1
1
693
XM_005270089.1
XP_005270146.1
1
693
XM_005270091.2
XP_005270148.1
1
693
XM_005270092.1
XP_005270149.1
1
693
XM_005270093.2
XP_005270150.1
1
693
XM_005270094.2
XP_005270151.1
1
693
XM_005270095.1
XP_005270152.1
1
693
XM_005270096.2
XP_005270153.1
1
693
XM_005270100.1
XP_005270157.1
1
693
XM_005270101.2
XP_005270158.1
1
693
XM_005270102.1
XP_005270159.1
1
693
XM_005270103.1
XP_005270160.1
1
693
XM_005270104.1
XP_005270161.1
1
693
XM_011540109.1
XP_011538411.1
1
693
XM_011540110.1
XP_011538412.1
1
693
XM_011540111.1
XP_011538413.1
1
693
XM_011540113.2
XP_011538415.1
1
693
XM_011540116.1
XP_011538418.1
1
693
XM_017016584.1
XP_016872073.1
1
693
XM_017016585.1
XP_016872074.1
1
693
XM_017016586.1
XP_016872075.1
1
693
XM_017016587.1
XP_016872076.1
1
693
XM_017016588.1
XP_016872077.1
1
693
XM_017016589.1
XP_016872078.1
1
693
XM_017016590.1
XP_016872079.1
1
693
XM_017016591.1
XP_016872080.1
1
693
XM_017016592.1
XP_016872081.1
1
693
XM_017016593.1
XP_016872082.1
1
693
XM_017016594.1
XP_016872083.1
1
693
XM_017016595.1
XP_016872084.1
1
693
XM_017016596.1
XP_016872085.1
1
693
GenBank mRNA
AB440195.1
-
1
155
AB451266.1
-
1
155
AK225809.1
-
1
666
AK299295.1
-
1
192
AK315243.1
-
1
328
BC032656.1
-
1
251
5533185
BX648364.1
-
1
30
CR536574.1
-
1
155
FJ010166.1
-
1
248
FJ010167.1
-
1
248
FJ010168.1
-
1
252
FJ010169.1
-
1
252
FJ010170.1
-
1
248
FJ010171.1
-
1
192
FJ010172.1
-
1
252
FJ010174.1
-
1
610
HM352838.1
-
1
155
HM352839.1
-
1
155
HM352840.1
-
1
155
HM352841.1
-
1
155
HM352842.1
-
1
155
HM352843.1
-
1
155
HM352844.1
-
1
155
HM352845.1
-
1
155
HM352846.1
-
1
155
HM352847.1
-
1
155
HM352848.1
-
1
155
HM352849.1
-
1
155
HM352850.1
-
1
155
HM352851.1
-
1
155
Y11306.2
-
1
462
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
blood vessel development
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
cell cycle arrest
Wnt signaling pathway, calcium modulating pathway
cell proliferation
response to glucose
positive regulation of heparan sulfate proteoglycan biosynthetic process
pancreas development
positive regulation of insulin secretion
positive regulation of protein binding
regulation of hormone metabolic process
glucose homeostasis
negative regulation of sequence-specific DNA binding transcription factor activity
maintenance of DNA repeat elements
canonical Wnt signaling pathway involved in positive regulation of epithelial to mesenchymal transition
fat cell differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of protein export from nucleus
myoblast fate commitment
regulation of smooth muscle cell proliferation
positive regulation of protein kinase B signaling
canonical Wnt signaling pathway
negative regulation of canonical Wnt signaling pathway
beta-catenin-TCF complex assembly
negative regulation of type B pancreatic cell apoptotic process
negative regulation of extrinsic apoptotic signaling pathway
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II repressing transcription factor binding
transcription factor activity, sequence-specific DNA binding
protein binding
beta-catenin binding
transcription factor binding
protein kinase binding
nuclear hormone receptor binding
sequence-specific DNA binding
transcription regulatory region DNA binding
gamma-catenin binding
armadillo repeat domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0