Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130413.1 | NP_569097.1 | 11, 12 | 1121 | ||
XM_006236477.2 | XP_006236539.1 | 11, 12 | 1106 | |||
XM_006236478.2 | XP_006236540.1 | 11, 12 | 1106 | |||
GenBank mRNA | AF302132.1 | - | 11, 12 | 1121 | ||
BC070949.1 | - | 11, 12 | 1068 | 7099436 |