Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Enterprise Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TCOF1 SIRNA Assays ›
Gene Symbol:
TCOF1
Gene Name:
treacle ribosome biogenesis factor 1
Gene Aliases:
MFD1, TCS, TCS1, treacle
Chromosome Location:
Chr.5: 150357639 - 150400308 on Build GRCh38
UniGene ID:
Hs.519672
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s13921
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TCOF1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000356.3
NP_000347.2
5
597
NM_001008657.2
NP_001008657.1
5
597
NM_001135243.1
NP_001128715.1
5
597
NM_001135244.1
NP_001128716.1
5
597
NM_001135245.1
NP_001128717.1
5
597
NM_001195141.1
NP_001182070.1
5
597
XM_005268502.3
XP_005268559.1
5
534
XM_005268503.3
XP_005268560.1
5
534
XM_005268504.3
XP_005268561.1
5
534
XM_005268505.3
XP_005268562.1
5
534
XM_005268506.3
XP_005268563.1
5
534
XM_005268507.3
XP_005268564.1
5
534
XM_005268508.3
XP_005268565.1
5
534
XM_005268509.3
XP_005268566.1
5
534
XM_011537678.2
XP_011535980.1
5
534
XM_017009792.1
XP_016865281.1
5
534
XM_017009793.1
XP_016865282.1
5
534
XM_017009794.1
XP_016865283.1
5
534
XM_017009795.1
XP_016865284.1
5
534
GenBank mRNA
AK096467.1
-
1
103
AK225284.1
-
5
522
AK303611.1
-
5
522
AK307586.1
-
5
522
AY460334.1
-
5
489
BC011764.2
-
5
518
3616898
BC014559.2
-
5
518
4025673
BC016144.1
-
5
517
BC027252.1
-
5
445
BC033093.1
-
5
518
4577077
BX537490.1
-
1
1254
U40847.1
-
5
582
U76366.1
-
5
564
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
skeletal system development
transport
transporter activity
protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0