Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130429.1 | NP_569113.1 | 9 | 1099 | ||
XM_006233302.2 | XP_006233364.1 | 9 | 2418 | |||
XM_006233303.2 | XP_006233365.1 | 9 | 2417 | |||
XM_006233305.2 | XP_006233367.1 | 8 | 2334 | |||
XM_006233306.2 | XP_006233368.1 | 8 | 2333 | |||
XM_006233309.2 | XP_006233371.1 | 9 | 1851 | |||
XM_008761489.1 | XP_008759711.1 | 9 | 2420 | |||
XM_008761490.1 | XP_008759712.1 | 8 | 2336 | |||
XM_008761491.1 | XP_008759713.1 | 9 | 1114 | |||
XM_008761492.1 | XP_008759714.1 | 9 | 1851 | |||
GenBank mRNA | AF198533.1 | - | 9 | 1099 |