siRNA Details
Gene Symbol: Sgk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134463.1 | NP_604458.1 | 13 | 1411 | ||
GenBank mRNA | AF361756.1 | - | 10 | 824 |

More Information
Other Species Targeted

Panther Classification:
