Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other AKR1C3 SIRNA Assays ›
          Gene Symbol:
          Akr1c3
          Gene Name:
          aldo-keto reductase family 1, member C3
          Gene Aliases:
          Akr1c18
          Chromosome Location:
          Chr.17: 69761126 - 69778021 on Build Rnor_6.0
          UniGene ID:
          Rn.10030
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID s140184

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Akr1c3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_138510.1 NP_612519.1 2 271
          GenBank mRNA BC086579.1 - 2 270
          BC100248.1 - 2 265 7307214
          D14424.1 - 2 256
          L32601.1 - 2 271

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_134066, BC034259, AK153898, AB059565.

          Panther Classification:

          Molecular Function -

          oxidoreductase reductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          protein import into nucleus, translocation
          prostaglandin metabolic process
          progesterone catabolic process
          G-protein coupled receptor signaling pathway
          parturition
          response to nutrient
          steroid metabolic process
          positive regulation of cell proliferation
          male gonad development
          cellular response to starvation
          positive regulation of cell death
          farnesol catabolic process
          keratinocyte differentiation
          cellular response to reactive oxygen species
          progesterone metabolic process
          retinal metabolic process
          multicellular organismal macromolecule metabolic process
          daunorubicin metabolic process
          doxorubicin metabolic process
          regulation of retinoic acid receptor signaling pathway
          regulation of steroid biosynthetic process
          positive regulation of protein kinase B signaling
          oxidation-reduction process
          testosterone biosynthetic process
          cellular response to cadmium ion
          cellular response to calcium ion
          cellular response to prostaglandin stimulus
          cellular response to corticosteroid stimulus
          cellular response to jasmonic acid stimulus
          cellular response to prostaglandin D stimulus
          negative regulation of retinoic acid biosynthetic process
          regulation of testosterone biosynthetic process
          positive regulation of endothelial cell apoptotic process
          positive regulation of reactive oxygen species metabolic process
          retinal dehydrogenase activity
          alditol:NADP+ 1-oxidoreductase activity
          retinol dehydrogenase activity
          oxidoreductase activity, acting on NAD(P)H, quinone or similar compound as acceptor
          phenanthrene 9,10-monooxygenase activity
          dihydrotestosterone 17-beta-dehydrogenase activity
          geranylgeranyl reductase activity
          ketoreductase activity
          17-alpha,20-alpha-dihydroxypregn-4-en-3-one dehydrogenase activity
          15-hydroxyprostaglandin-D dehydrogenase (NADP+) activity
          androsterone dehydrogenase activity
          ketosteroid monooxygenase activity
          delta4-3-oxosteroid 5beta-reductase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-5btgp:80/100.66.77.235:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0