Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFDP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007111.4 | NP_009042.1 | 9 | 1020 | ||
NR_026580.1 | 8 | 793 | ||||
XM_005268327.1 | XP_005268384.1 | 9 | 905 | |||
XM_005268328.2 | XP_005268385.1 | 9 | 958 | |||
XM_005268331.1 | XP_005268388.1 | 8 | 900 | |||
XM_011537518.2 | XP_011535820.1 | 8 | 872 | |||
XM_011537519.1 | XP_011535821.1 | 8 | 900 | |||
XM_011537520.2 | XP_011535822.1 | 8 | 783 | |||
XM_017020717.1 | XP_016876206.1 | 9 | 905 | |||
XM_017020718.1 | XP_016876207.1 | 9 | 905 | |||
XM_017020719.1 | XP_016876208.1 | 9 | 905 | |||
XM_017020720.1 | XP_016876209.1 | 8 | 866 | |||
XM_017020721.1 | XP_016876210.1 | 8 | 788 | |||
XM_017020722.1 | XP_016876211.1 | 8 | 783 | |||
GenBank mRNA | AK297114.1 | - | 8 | 783 | ||
AK298521.1 | - | 8 | 905 | |||
AK313742.1 | - | 8 | 866 | |||
BC011685.2 | - | 9 | 1020 | 4111289 | ||
L23959.1 | - | 8 | 845 |