Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Haus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138864.2 | NP_620219.2 | 2 | 209 | ||
GenBank mRNA | AF092207.1 | - | 205 | |||
BC127484.1 | - | 2 | 209 | 8363325 |
Gene Symbol: NEWGENE_619909 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772211.1 | XP_008770433.1 | 234 |