Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGFB1I1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042454.2 | NP_001035919.1 | 3, 4 | 213 | ||
NM_001164719.1 | NP_001158191.1 | 3, 4 | 182 | |||
NM_015927.4 | NP_057011.2 | 3, 4 | 212 | |||
GenBank mRNA | AB007836.1 | - | 2, 3 | 177 | ||
AF116343.1 | - | 2, 3 | 128 | |||
AK313327.1 | - | 3, 4 | 213 | |||
BC001507.3 | - | 3, 4 | 212 | 3030477 | ||
BC001830.2 | - | 3, 4 | 212 | 3030477 | ||
BC017288.2 | - | 3, 4 | 182 | 4901903 | ||
BC032545.1 | - | 2, 3 | 184 | 5535913 | ||
DA097743.1 | - | 3, 4 | 309 |