siRNA Details
Gene Symbol: Pcdhga4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772071.1 | XP_008770293.1 | 1 | 1890 |

Gene Symbol: Pcdhga4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772071.1 | XP_008770293.1 | 1 | 1890 |