Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Art5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013039.1 | NP_001013057.1 | 5 | 1154 | ||
XM_006229843.2 | XP_006229905.1 | 3 | 740 | |||
XM_006229845.2 | XP_006229907.1 | 3 | 742 | |||
XM_006229846.2 | XP_006229908.1 | 4 | 772 | |||
XM_008759692.1 | XP_008757914.1 | 3 | 684 | |||
XM_008759693.1 | XP_008757915.1 | 3 | 740 | |||
GenBank mRNA | AJ294469.1 | - | 1 | 198 | ||
BC083915.1 | - | 5 | 1154 | 7134556 |