Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003254.2 | NP_003245.1 | 4 | 500 | ||
XM_017029766.1 | XP_016885255.1 | 4 | 454 | |||
GenBank mRNA | A10416.1 | - | 4 | 370 | ||
AK074854.1 | - | 4 | 478 | |||
AK296116.1 | - | 3 | 643 | |||
AK310206.1 | - | 4 | 352 | |||
AK311937.1 | - | 4 | 352 | |||
BC000866.1 | - | 4 | 453 | 3460153 | ||
BC007097.1 | - | 4 | 401 | |||
BU857950.1 | - | 547 | ||||
CR407638.1 | - | 3 | 308 | |||
CR541982.1 | - | 3 | 308 | |||
M12670.1 | - | 4 | 356 | |||
M59906.1 | - | 4 | 351 | |||
S68252.1 | - | 3 | 311 | |||
X02598.1 | - | 4 | 349 | |||
X03124.1 | - | 4 | 370 |