Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006236922.2 | XP_006236984.1 | 12 | 2878 | ||
XM_006236923.2 | XP_006236985.1 | 12 | 2690 | |||
XM_006236924.2 | XP_006236986.1 | 11 | 2737 | |||
XM_008763139.1 | XP_008761361.1 | 12 | 2624 |