Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177927.2 | NP_808788.1 | 8 | 1086 | ||
GenBank mRNA | AY216659.1 | - | 8 | 1086 | ||
BC078686.1 | - | 8 | 1074 | 7110230 | ||
FQ218413.1 | - | 8 | 1087 | |||
FQ219756.1 | - | 8 | 1086 | |||
FQ220670.1 | - | 2 | 221 |