Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768160.1 | XP_008766382.1 | 12 | 1542 | ||
XM_008768161.1 | XP_008766383.1 | 12 | 1781 | |||
XM_008768162.1 | XP_008766384.1 | 12 | 1542 | |||
XM_008768163.1 | XP_008766385.1 | 12 | 1542 | |||
XM_008768164.1 | XP_008766386.1 | 11 | 1645 |