Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Supt4h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105828.2 | NP_001099298.2 | 3, 4 | 250 | ||
GenBank mRNA | BC168730.1 | - | 3, 4 | 250 | 6887426 | |
FQ217757.1 | - | 3, 4 | 257 | |||
FQ224188.1 | - | 3, 4 | 246 | |||
FQ228457.1 | - | 3, 4 | 244 |