Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TNNT3 SIRNA Assays ›
Gene Symbol:
TNNT3
Gene Name:
troponin T3, fast skeletal type
Gene Aliases:
TNTF
Chromosome Location:
Chr.11: 1919551 - 1938706 on Build GRCh38
UniGene ID:
Hs.73454
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s14287
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TNNT3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042780.2
NP_001036245.1
10
516
NM_001042781.2
NP_001036246.1
11
534
NM_001042782.2
NP_001036247.1
10
516
NM_001297646.1
NP_001284575.1
11
774
NM_006757.3
NP_006748.1
11
540
XM_006718288.3
XP_006718351.1
6
567
XM_006718290.3
XP_006718353.1
13
479
XM_006718293.2
XP_006718356.1
12
556
XM_006718294.3
XP_006718357.1
12
556
XM_006718296.3
XP_006718359.1
11
443
XM_006718299.2
XP_006718362.1
10
379
XM_006718300.3
XP_006718363.1
9
407
XM_006718302.3
XP_006718365.1
8
389
XM_011520343.2
XP_011518645.1
13
610
XM_017018205.1
XP_016873694.1
11
534
XM_017018206.1
XP_016873695.1
13
402
XM_017018207.1
XP_016873696.1
12
390
XM_017018208.1
XP_016873697.1
11
357
XM_017018209.1
XP_016873698.1
11
366
XM_017018210.1
XP_016873699.1
10
331
XM_017018211.1
XP_016873700.1
10
350
XM_017018212.1
XP_016873701.1
9
322
XM_017018213.1
XP_016873702.1
8
307
XM_017018214.1
XP_016873703.1
8
306
XM_017018215.1
XP_016873704.1
8
298
XM_017018216.1
XP_016873705.1
12
461
XM_017018217.1
XP_016873706.1
11
440
XM_017018218.1
XP_016873707.1
10
428
XM_017018219.1
XP_016873708.1
8
389
GenBank mRNA
AK056968.1
-
10
595
BC050446.1
-
10
372
BC117327.1
-
9
400
40125878
BC143537.1
-
9
400
9052047
BC171727.1
-
10
418
9052046
BC171728.1
-
6
353
BT019997.1
-
10
322
BX640689.1
-
11
398
CR541927.1
-
10
322
DA897990.1
-
11
534
DA899275.1
-
9
425
DQ778624.1
-
6
304
M21984.1
-
10
340
Back To Top
More Information
Panther Classification:
Molecular Function -
actin binding motor protein
Gene Ontology Categories:
Function(s)
Process(es)
skeletal muscle contraction
regulation of striated muscle contraction
muscle filament sliding
regulation of ATPase activity
actin binding
tropomyosin binding
troponin C binding
calcium-dependent ATPase activity
troponin I binding
calcium-dependent protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-mc8hj:80/100.66.75.242:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0