Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TNNT3 SIRNA Assays ›
Gene Symbol:
TNNT3
Gene Name:
troponin T3, fast skeletal type
Gene Aliases:
TNTF
Chromosome Location:
Chr.11: 1919551 - 1938706 on Build GRCh38
UniGene ID:
Hs.73454
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s14288
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TNNT3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042780.2
NP_001036245.1
12
751
NM_001042781.2
NP_001036246.1
13
769
NM_001042782.2
NP_001036247.1
12
751
NM_001297646.1
NP_001284575.1
13
1009
NM_006757.3
NP_006748.1
13
775
XM_006718288.3
XP_006718351.1
8
802
XM_006718290.3
XP_006718353.1
15
714
XM_006718293.2
XP_006718356.1
14
791
XM_006718294.3
XP_006718357.1
14
791
XM_006718296.3
XP_006718359.1
13
678
XM_006718299.2
XP_006718362.1
12
614
XM_006718300.3
XP_006718363.1
11
642
XM_006718302.3
XP_006718365.1
10
624
XM_011520343.2
XP_011518645.1
15
845
XM_017018205.1
XP_016873694.1
13
769
XM_017018206.1
XP_016873695.1
15
637
XM_017018207.1
XP_016873696.1
14
625
XM_017018208.1
XP_016873697.1
13
592
XM_017018209.1
XP_016873698.1
13
601
XM_017018210.1
XP_016873699.1
12
566
XM_017018211.1
XP_016873700.1
12
585
XM_017018212.1
XP_016873701.1
11
557
XM_017018213.1
XP_016873702.1
10
542
XM_017018214.1
XP_016873703.1
10
541
XM_017018215.1
XP_016873704.1
10
533
XM_017018216.1
XP_016873705.1
14
696
XM_017018217.1
XP_016873706.1
13
675
XM_017018218.1
XP_016873707.1
12
663
XM_017018219.1
XP_016873708.1
10
624
GenBank mRNA
BC050446.1
-
12
607
BC062430.1
-
9
333
BC117327.1
-
11
635
40125878
BC143537.1
-
11
635
9052047
BC171727.1
-
12
653
9052046
BC171728.1
-
8
588
BT019997.1
-
12
557
BX640689.1
-
13
633
CR541927.1
-
12
557
DQ778624.1
-
8
539
M21984.1
-
12
575
Back To Top
More Information
Panther Classification:
Molecular Function -
actin binding motor protein
Gene Ontology Categories:
Function(s)
Process(es)
skeletal muscle contraction
regulation of striated muscle contraction
muscle filament sliding
regulation of ATPase activity
actin binding
tropomyosin binding
troponin C binding
calcium-dependent ATPase activity
troponin I binding
calcium-dependent protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline