siRNA Details
Gene Symbol: TPBG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166392.1 | NP_001159864.1 | 2 | 1621 | ||
NM_006670.4 | NP_006661.1 | 3 | 2132 | |||
XM_011536097.2 | XP_011534399.1 | 2 | 1822 | |||
GenBank mRNA | AK074786.1 | - | 2 | 1619 | ||
AK074790.1 | - | 2 | 1621 | |||
AK291170.1 | - | 3 | 2132 | |||
BC037161.2 | - | 2 | 1622 | 4138906 | ||
Z29083.1 | - | 1 | 1280 |

More Information
Other Species Targeted

Panther Classification:
