Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other APC SIRNA Assays ›
          Gene Symbol:
          APC
          Gene Name:
          APC, WNT signaling pathway regulator
          Gene Aliases:
          BTPS2, DP2, DP2.5, DP3, GS, PPP1R46
          Chromosome Location:
          Chr.5: 112707505 - 112846239 on Build GRCh38
          UniGene ID:
          Hs.158932
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s1433
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 7,898.00
          Your Price
          Online offer:
          7898.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: APC
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000038.5 NP_000029.2 4 394
          NM_001127510.2 NP_001120982.1 5 502
          NM_001127511.2 NP_001120983.2 3 552
          GenBank mRNA AB210001.1 - 5 466
          AK289567.1 - 4 456
          AK294544.1 - 3 536
          BC056268.1 - 4 365
          BP230350.1 - 3 551
          DA795663.1 - 4 394
          M73548.1 - 3 347
          M74088.1 - 3 327

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_012499, XM_006254509, XM_006254511, XM_006254513, XM_006254520, and others.

          Matching Taqman Gene Expression Assay:


          Hs00181051_m1

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cytokinesis
          cell fate specification
          protein complex assembly
          cellular component disassembly involved in execution phase of apoptosis
          cellular response to DNA damage stimulus
          negative regulation of microtubule depolymerization
          cell cycle arrest
          mitotic spindle assembly checkpoint
          cell adhesion
          establishment or maintenance of cell polarity
          pattern specification process
          negative regulation of cell proliferation
          Wnt signaling pathway
          cell migration
          positive regulation of cell migration
          pancreas development
          neuron projection development
          positive regulation of pseudopodium assembly
          regulation of microtubule-based process
          response to drug
          positive regulation of apoptotic process
          proteasome-mediated ubiquitin-dependent protein catabolic process
          regulation of cell differentiation
          positive regulation of protein catabolic process
          negative regulation of cyclin-dependent protein serine/threonine kinase activity
          regulation of attachment of spindle microtubules to kinetochore
          canonical Wnt signaling pathway
          bicellular tight junction assembly
          negative regulation of canonical Wnt signaling pathway
          response to alcohol
          beta-catenin destruction complex assembly
          beta-catenin destruction complex disassembly
          cellular response to nerve growth factor stimulus
          regulation of glutamate metabolic process
          protein binding
          beta-catenin binding
          microtubule binding
          protein kinase regulator activity
          protein kinase binding
          protein complex binding
          gamma-catenin binding
          cadherin binding
          microtubule plus-end binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0