Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TPM3 SIRNA Assays ›
          Gene Symbol:
          TPM3 TPM3P9
          Gene Name:
          tropomyosin 3
          tropomyosin 3 pseudogene 9
          Gene Aliases:
          CAPM1, CFTD, HEL-189, HEL-S-82p, NEM1, OK/SW-cl.5, TM-5, TM3, TM30, TM30nm, TM5, TPMsk3, TRK, hscp30
          -
          Chromosome Location:
          Chr.1: 154155304 - 154192135 on Build GRCh38
          Chromosome Location:
          Chr.19: 53431974 - 53444672 on Build GRCh38
          UniGene ID:
          Hs.535581
          Hs.433293
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s14343

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TPM3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001043351.1 NP_001036816.1 4 526
          NM_001043352.1 NP_001036817.1 4 526
          NM_001043353.1 NP_001036818.1 4 526
          NM_001278188.1 NP_001265117.1 3 392
          NM_001278189.1 NP_001265118.1 4 526
          NM_001278190.1 NP_001265119.1 4 526
          NM_001278191.1 NP_001265120.1 4 372
          NM_152263.3 NP_689476.2 5 624
          NM_153649.3 NP_705935.1 4 526
          NR_103461.1 4 526
          GenBank mRNA AF474157.1 - 4 396
          AK026559.1 - 4 466
          AK056921.1 - 4 383
          AK056997.1 - 5 414
          AK092690.1 - 5 464
          AK092712.1 - 3 271
          AK125367.1 - 3 290
          AK298678.1 - 3 370
          AK301674.1 - 4 348
          AK308007.1 - 3 301
          AK315504.1 - 4 485
          AL537561.3 - 4 436
          AY004867.1 - 4 407
          BC000771.2 - 4 438 3506357
          BC008407.1 - 5 590 4245271
          BC008425.1 - 5 593 4246114
          BC015403.1 - 4 441 4389911
          BC062740.1 - 5 456 4271142
          BC072428.1 - 4 453 6472532
          BF967161.1 - 4 339
          BG706843.1 - 4 469
          BI546300.1 - 4 437
          BQ933569.1 - 4 493
          BU956481.1 - 4 452
          BX419658.2 - 4 439
          BX648485.1 - 4 438
          CR997359.1 - 4 417
          DA209890.1 - 4 375
          DA472065.1 - 4 526
          DA894064.1 - 5 414
          EU668324.1 - 5 464
          FJ224335.1 - 4 438
          KU178368.1 - 4 396
          KU178369.1 - 4 396
          KU178370.1 - 4 396
          M12128.1 - 1 24
          X04201.1 - 5 565
          X04588.1 - 4 448
          Gene Symbol: TPM3P9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003148.3 2 516
          GenBank mRNA BX647962.1 - 2 503

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001301286, NM_057208, L24777, L24776, L24775.

          Panther Classification:

          Molecular Function -

          actin binding motor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          movement of cell or subcellular component
          muscle contraction
          muscle filament sliding
          molecular_function
          actin binding
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0