Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grid2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105910.1 | NP_001099380.1 | 2, 3 | 325 | ||
XM_006248865.2 | XP_006248927.1 | 2, 3 | 395 | |||
XM_006248866.2 | XP_006248928.1 | 3, 4 | 987 |