Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TPR SIRNA Assays ›
Gene Symbol:
TPR
Gene Name:
translocated promoter region, nuclear basket protein
Gene Aliases:
-
Chromosome Location:
Chr.1: 186311652 - 186375325 on Build GRCh38
UniGene ID:
Hs.279640
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s14354
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TPR
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_003292.2
NP_003283.2
30
4389
GenBank mRNA
AK023111.1
-
5
2631
U69668.1
-
30
4180
X66397.1
-
30
4389
Back To Top
More Information
Panther Classification:
Molecular Function -
primary active transporter
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
MAPK import into nucleus
RNA import into nucleus
RNA export from nucleus
mRNA export from nucleus
tRNA export from nucleus
protein import into nucleus
protein export from nucleus
nuclear pore organization
mitotic nuclear division
mitotic nuclear envelope disassembly
mitotic spindle assembly checkpoint
biological_process
regulation of mRNA export from nucleus
regulation of glucose transport
regulation of mitotic sister chromatid separation
viral process
protein sumoylation
viral transcription
gene silencing by RNA
positive regulation of heterochromatin assembly
regulation of protein stability
mRNA export from nucleus in response to heat stress
cellular response to heat
cellular response to interferon-alpha
regulation of protein import into nucleus
positive regulation of protein import into nucleus
nuclear matrix organization
negative regulation of translational initiation
regulation of protein export from nucleus
positive regulation of protein export from nucleus
negative regulation of RNA export from nucleus
nuclear pore complex assembly
cell division
response to epidermal growth factor
intracellular transport of virus
positive regulation of mitotic cell cycle spindle assembly checkpoint
positive regulation of intracellular protein transport
regulation of cellular response to heat
regulation of mitotic spindle assembly
molecular_function
chromatin binding
mRNA binding
transporter activity
nucleocytoplasmic transporter activity
protein binding
tubulin binding
heat shock protein binding
protein homodimerization activity
protein anchor
poly(A) RNA binding
mitogen-activated protein kinase binding
dynein complex binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0