Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asmtl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105915.2 | NP_001099385.2 | 5 | 440 | ||
XM_006249001.2 | XP_006249063.1 | 6 | 557 | |||
GenBank mRNA | BC160824.1 | - | 4 | 781 | ||
FM053536.1 | - | 3 | 109 | |||
FM067394.1 | - | 5 | 447 |