Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000549.4 | NP_000540.2 | 3 | 325 | ||
NM_001277991.1 | NP_001264920.1 | 1 | 152 | |||
XM_011542065.2 | XP_011540367.1 | 2 | 364 | |||
GenBank mRNA | BC069298.1 | - | 2 | 287 | 7262720 |