Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TTC3 SIRNA Assays ›
Gene Symbol:
TTC3
Gene Name:
tetratricopeptide repeat domain 3
Gene Aliases:
DCRR1, RNF105, TPRDIII
Chromosome Location:
Chr.21: 37073184 - 37203118 on Build GRCh38
UniGene ID:
Hs.368214
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s14476
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TTC3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001001894.1
NP_001001894.1
33
3887
NM_001320703.1
NP_001307632.1
34
4147
NM_001320704.1
NP_001307633.1
34
4137
NM_003316.3
NP_003307.3
33
5178
XM_005261048.1
XP_005261105.1
35
4047
XM_005261049.1
XP_005261106.1
35
3970
XM_005261050.4
XP_005261107.1
35
6932
XM_005261052.1
XP_005261109.1
34
3922
XM_005261054.3
XP_005261111.1
34
6876
XM_005261055.1
XP_005261112.1
25
3102
XM_005261056.2
XP_005261113.1
25
3181
XM_011529739.2
XP_011528041.1
27
3253
XM_011529740.2
XP_011528042.1
31
3707
XM_017028458.1
XP_016883947.1
35
3986
XM_017028459.1
XP_016883948.1
34
3931
XM_017028460.1
XP_016883949.1
33
3882
XM_017028461.1
XP_016883950.1
34
3993
XM_017028462.1
XP_016883951.1
34
3916
XM_017028463.1
XP_016883952.1
33
3945
XM_017028464.1
XP_016883953.1
25
3221
XM_017028465.1
XP_016883954.1
31
5332
XM_017028466.1
XP_016883955.1
24
3143
XM_017028467.1
XP_016883956.1
24
3048
GenBank mRNA
AY523634.1
-
6
956
BC137345.1
-
33
4317
9021343
CR936616.1
-
18
2483
D83077.1
-
33
5178
D83327.1
-
30
3530
D84294.1
-
32
5251
D84295.1
-
31
5165
D84296.1
-
23
3589
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
ubiquitin-dependent protein catabolic process
protein K48-linked ubiquitination
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-kz7s5:80/10.239.165.170:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0