Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HNRNPC SIRNA Assays ›
          Gene Symbol:
          Hnrnpc LOC100911576
          Gene Name:
          heterogeneous nuclear ribonucleoprotein C
          heterogeneous nuclear ribonucleoproteins C1/C2-like
          Gene Aliases:
          Hnrpc, hnRNP C
          -
          Chromosome Location:
          Chr.15: 28486605 - 28517353 on Build Rnor_6.0
          Chromosome Location:
          Chr.14: 88543630 - 88574207 on Build Rnor_6.0
          UniGene ID:
          Rn.101929
          Rn.101929
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s144763

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Hnrnpc
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001277600.1 NP_001264529.1 3 381
          NM_001277601.1 NP_001264530.1 3 381
          NM_001277602.1 NP_001264531.1 2 355
          XM_006251858.2 XP_006251920.1 2 849
          XM_006251859.2 XP_006251921.1 2 342
          XM_006251861.2 XP_006251923.1 2 848
          XM_006251862.2 XP_006251924.1 4 532
          XM_006251863.2 XP_006251925.1 2 342
          XM_006251864.2 XP_006251926.1 4 437
          XM_006251865.2 XP_006251927.1 2 848
          XM_006251866.1 XP_006251928.1 3 368
          XM_008770508.1 XP_008768730.1 3 876
          XM_008770509.1 XP_008768731.1 3 1944
          XM_008770510.1 XP_008768732.1 3 376
          XM_008770511.1 XP_008768733.1 3 376
          XM_008770512.1 XP_008768734.1 3 368
          GenBank mRNA BC097346.1 - 2 305 7442283
          CB582273.1 - 3 331
          FQ222540.1 - 1 31
          FQ227266.1 - 1 31
          Gene Symbol: LOC100911576
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001271241.1 NP_001258170.1 3 346
          XM_003751394.2 XP_003751442.1 3 332
          XM_003751395.2 XP_003751443.1 3 332
          XM_006251449.2 XP_006251511.1 3 1943
          XM_006251450.2 XP_006251512.1 2 847
          XM_006251451.1 XP_006251513.1 2 306
          XM_006251452.2 XP_006251514.1 3 332
          XM_006251453.1 XP_006251515.1 4 496
          XM_006251454.1 XP_006251516.1 4 401
          XM_006251455.2 XP_006251517.1 2 846
          XM_006251456.1 XP_006251518.1 2 306
          XM_006251457.1 XP_006251519.1 2 306
          XM_006251458.2 XP_006251520.1 3 332
          XM_008770242.1 XP_008768464.1 3 840
          XM_008770243.1 XP_008768465.1 3 332
          GenBank mRNA AY724499.1 - 2 774
          FQ214198.1 - 1 31
          FQ215781.1 - 1 31
          FQ227893.1 - 1 31
          FQ228082.1 - 1 31

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_001170983, NM_001170984, NM_001170982, NM_001170981, NM_016884, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          mRNA processing
          RNA splicing
          ATP-dependent chromatin remodeling
          3'-UTR-mediated mRNA stabilization
          nucleotide binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II distal enhancer sequence-specific DNA binding
          nucleic acid binding
          RNA binding
          mRNA binding
          mRNA 3'-UTR binding
          poly(U) RNA binding
          nucleosomal DNA binding
          identical protein binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0