Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcpt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170466.1 | NP_001163937.1 | 2, 3 | 211 | ||
GenBank mRNA | U67888.1 | - | 2, 3 | 188 |
Gene Symbol: LOC103690000 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770718.1 | XP_008768940.1 | 190 |