Search
Search

Haus8| Gene Symbol: Haus8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024971.2 | NP_001020142.2 | 6 | 582 | ||
| XM_008771074.1 | XP_008769296.1 | 7 | 724 | |||
| XM_008771075.1 | XP_008769297.1 | 7 | 721 | |||
| XM_008771076.1 | XP_008769298.1 | 7 | 721 | |||
| XM_008771077.1 | XP_008769299.1 | 6 | 627 | |||
| XM_008771078.1 | XP_008769300.1 | 3 | 245 | |||
| GenBank mRNA | BC091198.1 | - | 6 | 571 | 7376974 | |
| DY317896.1 | - | 6 | 542 | |||
| FQ225627.1 | - | 6 | 594 | |||