Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYRO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006293.3 | NP_006284.2 | 19 | 2575 | ||
XM_017022542.1 | XP_016878031.1 | 19 | 2540 | |||
GenBank mRNA | AB209431.1 | - | 19 | 2604 | ||
AK131389.1 | - | 1 | 608 | |||
AK295496.1 | - | 8 | 870 | |||
BC029925.2 | - | 11 | 1225 | |||
BC034960.1 | - | 19 | 2357 | |||
BC049368.1 | - | 19 | 2349 | 4821587 | ||
BC051756.1 | - | 19 | 2354 | 6095596 | ||
D17517.1 | - | 19 | 2563 | |||
D50479.1 | - | 19 | 2335 | |||
U02566.1 | - | 16 | 1780 | |||
U05682.1 | - | 19 | 2345 | |||
X72886.1 | - | 8 | 785 |