Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myot | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106148.1 | NP_001099618.1 | 8 | 1314 | ||
XM_006254666.2 | XP_006254728.1 | 8 | 1214 | |||
XM_006254667.2 | XP_006254729.1 | 8 | 1331 | |||
XM_006254668.2 | XP_006254730.1 | 8 | 1211 | |||
GenBank mRNA | FQ215235.1 | - | 5 | 661 | ||
FQ215267.1 | - | 6 | 680 |
Gene Symbol: LOC100909761 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006254990.2 | XP_006255052.1 | 8 | 1226 | ||
XM_006254991.2 | XP_006255053.1 | 8 | 1343 | |||
XM_006254992.2 | XP_006255054.1 | 8 | 1223 |
Gene Symbol: LOC100909899 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006254989.2 | XP_006255051.1 | 698 | |||
XM_008772217.1 | XP_008770439.1 | 696 |
Gene Symbol: LOC100912306 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244679.2 | XP_006244741.1 | 8 | 1214 | ||
XM_006244680.2 | XP_006244742.1 | 8 | 1331 | |||
XM_006244681.2 | XP_006244743.1 | 8 | 1211 |
Gene Symbol: LOC100912348 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244682.2 | XP_006244744.1 | 699 | |||
XM_008766963.1 | XP_008765185.1 | 696 |