Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other UBE2V1 SIRNA Assays ›
          Gene Symbol:
          UBE2V1 TMEM189-UBE2V1
          Gene Name:
          ubiquitin conjugating enzyme E2 V1
          TMEM189-UBE2V1 readthrough
          Gene Aliases:
          CIR1, CROC-1, CROC1, UBE2V, UEV-1, UEV1, UEV1A
          CROC-1B, CROC1B, KUA-UEV
          Chromosome Location:
          Chr.20: 50081124 - 50115959 on Build GRCh38
          Chromosome Location:
          Chr.20: 50081124 - 50153798 on Build GRCh38
          UniGene ID:
          Hs.420529
          Hs.744839
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s14598

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: UBE2V1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001032288.2 NP_001027459.1 3 354
          NM_001257393.1 NP_001244322.1 4 529
          NM_001257394.1 NP_001244323.1 2 588
          NM_001257395.1 NP_001244324.1 3 345
          NM_001257397.1 NP_001244326.1 4 437
          NM_001257398.1 NP_001244327.1 4 493
          NM_001257399.1 NP_001244328.1 4 472
          NM_001282575.2 NP_001269504.1 4 789
          NM_001282576.1 NP_001269505.1 4 628
          NM_001282577.2 NP_001269506.1 4 406
          NM_001282578.2 NP_001269507.1 3 543
          NM_001282579.1 NP_001269508.1 4 454
          NM_021988.5 NP_068823.2 3 737
          NM_022442.5 NP_071887.1 3 380
          NM_199144.2 NP_954595.1 4 660
          NR_047554.1 2 205
          GenBank mRNA AK314754.1 - 3 725
          AL110132.1 - 2 1155
          AY008273.1 - 3 353
          BC000468.2 - 3 294 2961017
          BC005215.1 - 4 787
          BC008944.2 - 2 271
          BT007382.1 - 3 284
          BU540046.1 - 4 661
          BU595183.1 - 331
          BX647511.1 - 2 173
          BX648147.1 - 2 179
          CA453411.1 - 378
          CD103154.1 - 2 517
          CN346855.1 - 4 461
          DA087027.1 - 4 354
          DA519011.1 - 3 354
          DA573591.1 - 4 424
          DA625165.1 - 4 439
          DA642098.1 - 4 570
          DA885410.1 - 3 297
          DA892136.1 - 4 431
          DA916492.1 - 3 495
          DB448941.1 - 4 406
          HY019085.1 - 4 426
          U39360.1 - 4 422
          U97280.1 - 3 297
          U97281.1 - 2 204
          Gene Symbol: TMEM189-UBE2V1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_199203.2 NP_954673.1 7 1114
          GenBank mRNA BU156219.1 - 3 289
          BU619725.1 - 1 87
          CF145300.1 - 16
          U39361.1 - 4 575

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          stimulatory C-type lectin receptor signaling pathway
          regulation of DNA repair
          postreplication repair
          regulation of transcription, DNA-templated
          cell differentiation
          Fc-epsilon receptor signaling pathway
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of transcription, DNA-templated
          T cell receptor signaling pathway
          positive regulation of NF-kappaB transcription factor activity
          nucleotide-binding oligomerization domain containing signaling pathway
          protein K63-linked ubiquitination
          protein binding
          ubiquitin protein ligase binding
          ubiquitin protein ligase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-pq742:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0