siRNA Details
Gene Symbol: UBTF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076683.1 | NP_001070151.1 | 4 | 466 | ||
NM_001076684.2 | NP_001070152.1 | 4 | 552 | |||
NM_014233.3 | NP_055048.1 | 4 | 552 | |||
NR_045058.1 | 4 | 749 | ||||
XM_006722059.3 | XP_006722122.1 | 4 | 591 | |||
XM_006722060.2 | XP_006722123.1 | 3 | 335 | |||
XM_006722061.2 | XP_006722124.1 | 4 | 419 | |||
XM_017024995.1 | XP_016880484.1 | 4 | 600 | |||
XM_017024996.1 | XP_016880485.1 | 4 | 552 | |||
XM_017024997.1 | XP_016880486.1 | 4 | 744 | |||
XM_017024998.1 | XP_016880487.1 | 4 | 460 | |||
XM_017024999.1 | XP_016880488.1 | 3 | 335 | |||
XM_017025000.1 | XP_016880489.1 | 4 | 504 | |||
XM_017025001.1 | XP_016880490.1 | 4 | 552 | |||
XM_017025002.1 | XP_016880491.1 | 4 | 432 | |||
XM_017025003.1 | XP_016880492.1 | 4 | 442 | |||
XM_017025004.1 | XP_016880493.1 | 4 | 434 | |||
GenBank mRNA | AK291733.1 | - | 4 | 377 | ||
AK292518.1 | - | 4 | 749 | |||
AK292577.1 | - | 4 | 637 | |||
AK298008.1 | - | 2 | 78 | |||
AK308321.1 | - | 3 | 529 | |||
BC031423.1 | - | 3 | 490 | |||
BC042297.1 | - | 4 | 457 | 4509695 | ||
BP230502.1 | - | 4 | 466 | |||
X53390.1 | - | 3 | 403 | |||
X53461.1 | - | 3 | 403 | |||
X56687.1 | - | 4 | 435 |

More Information
Other Species Targeted

Panther Classification:
