Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001521.1 | NP_001001521.1 | 6 | 963 | ||
NM_006759.3 | NP_006750.3 | 6 | 1019 | |||
XM_005264537.2 | XP_005264594.1 | 7 | 1071 | |||
XM_005264538.1 | XP_005264595.1 | 7 | 1038 | |||
XM_006712088.1 | XP_006712151.1 | 7 | 1144 | |||
XM_011533091.1 | XP_011531393.1 | 4 | 698 | |||
XM_017004855.1 | XP_016860344.1 | 7 | 1039 | |||
XM_017004856.1 | XP_016860345.1 | 8 | 931 | |||
XM_017004857.1 | XP_016860346.1 | 8 | 1232 | |||
GenBank mRNA | AK300731.1 | - | 6 | 747 | ||
AK314263.1 | - | 6 | 1010 | |||
BC000173.2 | - | 6 | 964 | 2985723 | ||
BC002954.1 | - | 6 | 843 | 3542849 | ||
BC047004.1 | - | 6 | 970 | 3924393 | ||
BX537559.1 | - | 2 | 3810 | |||
BX647825.1 | - | 4 | 811 | |||
U27460.1 | - | 7 | 788 |