Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other UGT2B11 SIRNA Assays ›
          Gene Symbol:
          UGT2B11 UGT2B10 UGT2B28
          Gene Name:
          UDP glucuronosyltransferase family 2 member B11
          UDP glucuronosyltransferase family 2 member B10
          UDP glucuronosyltransferase family 2 member B28
          Gene Aliases:
          -
          UDPGT2B10
          -
          Chromosome Location:
          Chr.4: 69200317 - 69224983 on Build GRCh38
          Chromosome Location:
          Chr.4: 68815981 - 68832038 on Build GRCh38
          Chromosome Location:
          Chr.4: 69280429 - 69295050 on Build GRCh38
          UniGene ID:
          Hs.339811
          Hs.201634
          Hs.653154
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID s14656

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: UGT2B11
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_011531550.2 XP_011529852.1 2 571
          XM_017007660.1 XP_016863149.1 3 323
          XM_017007661.1 XP_016863150.1 2 201
          GenBank mRNA AF016492.1 - 2 817
          BC069441.1 - 2 817 7262335
          BC107059.2 - 2 817 40006084
          BC107060.2 - 2 817 40006085
          Gene Symbol: UGT2B10
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001075.5 NP_001066.1 2 832
          NM_001144767.2 NP_001138239.1 2 580
          NM_001290091.1 NP_001277020.1 2 267
          XM_017008585.1 XP_016864074.1 2 844
          GenBank mRNA AK222839.1 - 2 830
          AK222872.1 - 2 834
          AK292738.1 - 2 838
          AK298432.1 - 2 579
          BC113649.1 - 2 833 8322701
          CD013994.1 - 2 241
          Gene Symbol: UGT2B28
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001207004.1 NP_001193933.1 2 810
          NM_053039.1 NP_444267.1 2 810
          XM_017008303.1 XP_016863792.1 2 880
          GenBank mRNA AB451456.1 - 2 808
          AF177272.1 - 2 810
          AF177273.1 - 2 810
          AF177274.1 - 3 459

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          xenobiotic metabolic process
          estrogen metabolic process
          flavonoid biosynthetic process
          flavonoid glucuronidation
          lipid metabolic process
          metabolic process
          glucuronosyltransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-v7cl8:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0