Search
Search

Catsperg1| Gene Symbol: Catsperg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170340.1 | NP_001163811.1 | 19 | 2219 | ||
| XM_006228722.2 | XP_006228784.1 | 19 | 2805 | |||
| XM_008759124.1 | XP_008757346.1 | 19 | 2838 | |||
| XM_008759125.1 | XP_008757347.1 | 19 | 2820 | |||
| XM_008759126.1 | XP_008757348.1 | 18 | 2730 | |||
| XM_008759127.1 | XP_008757349.1 | 18 | 2036 | |||
| GenBank mRNA | BC167101.1 | - | 12 | 1232 | ||