Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tjp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106266.1 | NP_001099736.1 | 22 | 4031 | ||
XM_006229330.2 | XP_006229392.1 | 23 | 4174 | |||
XM_006229331.2 | XP_006229393.1 | 23 | 4263 | |||
XM_006229332.2 | XP_006229394.1 | 23 | 4553 | |||
XM_006229333.2 | XP_006229395.1 | 23 | 4073 | |||
XM_008759501.1 | XP_008757723.1 | 23 | 4497 | |||
XM_008759502.1 | XP_008757724.1 | 23 | 4497 | |||
XM_008759503.1 | XP_008757725.1 | 22 | 4257 | |||
XM_008759504.1 | XP_008757726.1 | 22 | 4257 |