Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170434.1 | NP_001163905.1 | 2, 3 | 149 | ||
XM_006229746.2 | XP_006229808.1 | 2, 3 | 999 | |||
XM_006229747.2 | XP_006229809.1 | 2, 3 | 1006 | |||
GenBank mRNA | FM029675.1 | - | 2, 3 | 149 |