Search
Search

| Gene Symbol: Olr168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000174.1 | NP_001000174.1 | 415 | |||
| Gene Symbol: Olr159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000170.1 | NP_001000170.1 | 1 | 415 | ||
| Gene Symbol: Olr163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000171.1 | NP_001000171.1 | 1 | 415 | ||