Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KDM6A SIRNA Assays ›
Gene Symbol:
KDM6A
Gene Name:
lysine demethylase 6A
Gene Aliases:
KABUK2, UTX, bA386N14.2
Chromosome Location:
Chr.X: 44873175 - 45112612 on Build GRCh38
UniGene ID:
Hs.522616
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s14737
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KDM6A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001291415.1
NP_001278344.1
22
3731
NM_001291416.1
NP_001278345.1
21
3596
NM_001291417.1
NP_001278346.1
20
3440
NM_001291418.1
NP_001278347.1
19
3338
NM_001291421.1
NP_001278350.1
19
3319
NM_021140.3
NP_066963.2
21
3575
NR_111960.1
20
3494
XM_005272656.4
XP_005272713.1
21
3629
XM_005272659.4
XP_005272716.1
20
3473
XM_011543957.2
XP_011542259.1
23
3788
XM_011543958.2
XP_011542260.1
22
3731
XM_011543959.2
XP_011542261.1
22
3686
XM_011543960.2
XP_011542262.1
23
3788
XM_011543961.2
XP_011542263.1
22
3653
XM_011543962.2
XP_011542264.1
22
3632
XM_011543963.2
XP_011542265.1
21
3596
XM_011543964.2
XP_011542266.1
21
3575
XM_011543965.2
XP_011542267.1
21
3551
XM_011543966.2
XP_011542268.1
21
3530
XM_011543967.2
XP_011542269.1
22
3632
XM_011543968.2
XP_011542270.1
21
3497
XM_011543969.2
XP_011542271.1
20
3494
XM_011543970.2
XP_011542272.1
20
3473
XM_011543971.2
XP_011542273.1
21
3551
XM_011543972.2
XP_011542274.1
20
3440
XM_011543973.2
XP_011542275.1
21
3497
XM_011543974.1
XP_011542276.1
21
3575
XM_011543975.2
XP_011542277.1
18
2901
XM_017029782.1
XP_016885271.1
22
3653
XM_017029783.1
XP_016885272.1
20
3494
XM_017029784.1
XP_016885273.1
20
3041
XM_017029785.1
XP_016885274.1
18
3014
GenBank mRNA
AB208795.1
-
21
3528
AF000992.1
-
21
3224
AF000993.1
-
21
3224
AK303430.1
-
10
2179
AL831996.2
-
10
2101
BC093868.1
-
21
3221
7939713
BC113381.1
-
21
3221
8322433
BC143272.1
-
21
3242
9051774
BC143277.1
-
22
3377
9051779
BC143278.1
-
20
3086
9051780
DQ062677.1
-
20
2961
DQ062678.1
-
20
3063
EU026283.1
-
21
3219
EU026291.1
-
20
3117
Back To Top
More Information
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
neural tube closure
heart morphogenesis
respiratory system process
positive regulation of gene expression
somite rostral/caudal axis specification
multicellular organism growth
mesodermal cell differentiation
notochord morphogenesis
histone H3-K4 methylation
oxidation-reduction process
canonical Wnt signaling pathway
histone H3-K27 demethylation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
chromatin DNA binding
histone demethylase activity
identical protein binding
metal ion binding
dioxygenase activity
histone demethylase activity (H3-K27 specific)
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-f6n66:80/10.239.161.116:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0