Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100519.1 | NP_001093989.1 | 17 | 1835 | ||
GenBank mRNA | BC098781.1 | - | 11 | 1113 | ||
FQ234875.1 | - | 4 | 431 |