Search
Search

VHL| Gene Symbol: VHL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000551.3 | NP_000542.1 | 3 | 770 | ||
| NM_198156.2 | NP_937799.1 | 2 | 647 | |||
| GenBank mRNA | AK304002.1 | - | 3 | 355 | ||
| AK309560.1 | - | 3 | 694 | |||
| AK315799.1 | - | 3 | 629 | |||
| BC058831.1 | - | 2 | 498 | 6457060 | ||
| KU178386.1 | - | 2 | 434 | |||
| KU178387.1 | - | 3 | 557 | |||
| L15409.1 | - | 3 | 770 | |||