Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xlr4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006229523.2 | XP_006229585.1 | 5, 6 | 907 | ||
XM_006229524.2 | XP_006229586.1 | 5, 6 | 922 | |||
XM_006229527.2 | XP_006229589.1 | 5, 6 | 527 | |||
XM_006229528.2 | XP_006229590.1 | 5, 6 | 889 | |||
XM_006229529.2 | XP_006229591.1 | 5, 6 | 904 | |||
XM_219738.8 | XP_219738.6 | 4, 5 | 1066 |