Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011559.1 | NP_001011559.1 | 12 | 1818 | ||
XM_006256248.2 | XP_006256310.1 | 13 | 2008 | |||
XM_006256249.2 | XP_006256311.1 | 13 | 2008 | |||
XM_008772811.1 | XP_008771033.1 | 13 | 2008 | |||
GenBank mRNA | AB193179.1 | - | 12 | 1849 | ||
AY749166.1 | - | 12 | 1818 |