Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcm3ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106382.1 | NP_001099852.1 | 3 | 1745 | ||
XM_006256293.2 | XP_006256355.1 | 4 | 1927 | |||
XM_006256294.2 | XP_006256356.1 | 3 | 1855 |