Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naip5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760694.1 | XP_008758916.1 | 11 | 3036 | ||
XM_226743.6 | XP_226743.4 | 11 | 3020 |