Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other APOC3 SIRNA Assays ›
          Gene Symbol:
          APOC3
          Gene Name:
          apolipoprotein C3
          Gene Aliases:
          APOCIII, HALP2
          Chromosome Location:
          Chr.11: 116829908 - 116833071 on Build GRCh38
          UniGene ID:
          Hs.73849
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s1489

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: APOC3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000040.1 NP_000031.1 4 275
          GenBank mRNA BC027977.1 - 4 273 5229117
          BC121081.1 - 4 275 40119733
          BC134419.1 - 4 275 40119734
          M28613.1 - 3 259
          M28614.1 - 3 259
          X00567.1 - 3 229
          X01388.1 - 4 300

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          apolipoprotein

          Gene Ontology Categories:

          Function(s) Process(es)

          retinoid metabolic process
          triglyceride metabolic process
          inflammatory response
          G-protein coupled receptor signaling pathway
          positive regulation of triglyceride biosynthetic process
          negative regulation of triglyceride catabolic process
          negative regulation of very-low-density lipoprotein particle remodeling
          negative regulation of very-low-density lipoprotein particle clearance
          negative regulation of high-density lipoprotein particle clearance
          negative regulation of low-density lipoprotein particle clearance
          triglyceride catabolic process
          regulation of Cdc42 protein signal transduction
          response to vitamin A
          cholesterol efflux
          phospholipid efflux
          response to triglyceride
          high-density lipoprotein particle remodeling
          very-low-density lipoprotein particle assembly
          chylomicron remnant clearance
          lipoprotein metabolic process
          response to drug
          cholesterol homeostasis
          lipoprotein transport
          response to peptide hormone
          reverse cholesterol transport
          negative regulation of fatty acid biosynthetic process
          negative regulation of lipid metabolic process
          negative regulation of receptor-mediated endocytosis
          negative regulation of lipid catabolic process
          negative regulation of lipoprotein lipase activity
          negative regulation of cholesterol import
          triglyceride homeostasis
          response to fatty acid
          cellular response to glucose stimulus
          negative regulation of oxidative phosphorylation
          response to simvastatin
          phospholipid binding
          cholesterol binding
          enzyme regulator activity
          lipase inhibitor activity
          high-density lipoprotein particle receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0