Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other WNT11 SIRNA Assays ›
          Gene Symbol:
          WNT11
          Gene Name:
          Wnt family member 11
          Gene Aliases:
          HWNT11
          Chromosome Location:
          Chr.11: 76186326 - 76210842 on Build GRCh38
          UniGene ID:
          Hs.108219
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s14893

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: WNT11
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_004626.2 NP_004617.2 4 742
          XM_005274231.1 XP_005274288.1 5 998
          XM_011545238.1 XP_011543540.1 5 1028
          XM_011545239.2 XP_011543541.1 5 1236
          XM_011545240.2 XP_011543542.1 6 1192
          XM_011545241.2 XP_011543543.1 4 480
          XM_017018241.1 XP_016873730.1 3 1500
          XM_017018242.1 XP_016873731.1 3 1335
          XM_017018243.1 XP_016873732.1 2 154
          GenBank mRNA AB070218.1 - 4 681
          AK313570.1 - 4 744
          BC074790.2 - 4 661 30915564
          BC074791.2 - 4 661 30915347
          BC113386.1 - 4 662 8322438
          BC113388.1 - 4 662 8322440
          JQ715612.1 - 4 498
          Y12692.1 - 4 742

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          epithelial to mesenchymal transition
          outflow tract morphogenesis
          planar cell polarity pathway involved in axis elongation
          protein phosphorylation
          Wnt signaling pathway, calcium modulating pathway
          positive regulation of gene expression
          Wnt signaling pathway
          neuron differentiation
          bone mineralization
          negative regulation of cell growth
          adrenal gland development
          positive regulation of cell migration
          negative regulation of cell migration
          response to nutrient levels
          activation of protein kinase activity
          positive regulation of transforming growth factor beta2 production
          protein localization to cell surface
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          positive regulation of GTPase activity
          cell fate commitment
          maintenance of epithelial cell apical/basal polarity
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          paraxial mesoderm formation
          notochord morphogenesis
          embryonic skeletal system development
          artery morphogenesis
          positive regulation of stress fiber assembly
          palate development
          convergent extension involved in axis elongation
          canonical Wnt signaling pathway
          Wnt signaling pathway, planar cell polarity pathway
          cloacal septation
          ventricular septum morphogenesis
          lung-associated mesenchyme development
          negative regulation of cell death
          ureteric bud morphogenesis
          planar cell polarity pathway involved in gastrula mediolateral intercalation
          negative regulation of cartilage development
          somite development
          neuroendocrine cell differentiation
          bicellular tight junction assembly
          cellular response to retinoic acid
          mesonephric duct development
          negative regulation of mesenchymal cell proliferation
          positive regulation of protein kinase C signaling
          negative regulation of canonical Wnt signaling pathway
          negative regulation of fibroblast growth factor production
          GTPase activator activity
          frizzled binding
          protein binding
          protein kinase activator activity
          transcription regulatory region DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0