Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039196.1 | NP_001034285.1 | 6, 7 | 890 | ||
XM_006234501.2 | XP_006234563.1 | 6, 7 | 1096 | |||
XM_006234502.2 | XP_006234564.1 | 5, 6 | 998 | |||
GenBank mRNA | BC112321.1 | - | 6, 7 | 890 | 7933412 |